Anti-PARP1 Mouse Monoclonal Antibody [clone: 194C1439]
NOVUNBP2-27335
- Antibody type:Primary
- Antigen name:Poly (ADP-ribose) polymerase 1
- Antigen symbol:PARP1
- Clonality:Monoclonal
- Clone:194C1439
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Gene ID:142
- Antigen synonyms:PPOL|PARP-1|ADPRT1|pADPRT-1|ADPRT 1|PARP|ARTD1|ADPRT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: monoclonal
Clone: 194C1439
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human