Anti-TMEM26 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27334
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:transmembrane protein 26
- Antigen symbol:TMEM26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:219623
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TMEM26 (transmembrane protein 26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human