Order Entry
Germany
ContactUsLinkComponent
Anti-NR5A2/LRH1 Rabbit Polyclonal Antibody
Anti-NR5A2/LRH1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27204
Supplier:  Novus Biologicals
undefined
Anti-NR5A2/LRH1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27204
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    nuclear receptor subfamily 5, group A, member 2
  • Antigen symbol:
    NR5A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Gene ID:
    2494
  • Antigen synonyms:
    B1F|B1F2|FTZ-F1beta|LRH-1|FTZ-F1|hB1F-2|LRH1|FTF|CPF
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: NR5A2/LRH1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: