Anti-NEDD4 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27197
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Neural Precursor Cell Expressed, Developmentally Down-Regulated 4
- Antigen symbol:NEDD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:4734
- Antigen synonyms:RPF1|NEDD4-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NEDD4 (neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: