Anti-IMPDH2 Rabbit Polyclonal Antibody
NOVUNBP2-27195
- Antibody type:Primary
- Antigen name:IMP (inosine 5'-monophosphate) dehydrogenase 2
- Antigen symbol:IMPDH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:3615
- Antigen synonyms:IMPD2|IMPDH-II
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: IMPDH2 (IMP (inosine 5'-monophosphate) dehydrogenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: