Anti-TLR2 Rat Monoclonal Antibody [clone: 11G5]
NOVUNBP2-27166
- Antibody type:Primary
- Antigen name:Toll-like Receptor 2
- Antigen symbol:TLR2
- Clonality:Monoclonal
- Clone:11G5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Reactivity:Human
- Western blot:Yes
- Gene ID:7097
- Antigen synonyms:CD282|TIL4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: 11G5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human