Order Entry
Germany
ContactUsLinkComponent
Anti-Macrophage Inflammatory Protein 3 alpha Rat Monoclonal Antibody [clone: 4N5F7]
Anti-Macrophage Inflammatory Protein 3 alpha Rat Monoclonal Antibody [clone: 4N5F7]
Catalog # NOVUNBP2-27142
Supplier:  Novus Biologicals
undefined
Anti-Macrophage Inflammatory Protein 3 alpha Rat Monoclonal Antibody [clone: 4N5F7]
Catalog # NOVUNBP2-27142
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    chemokine (C-C motif) ligand 20
  • Antigen symbol:
    CCL20
  • Clonality:
    Monoclonal
  • Clone:
    4N5F7
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    6364
  • Antigen synonyms:
    Exodus|MIP3A|MIP-3-alpha|SCYA20|CKb4|MIP-3a|ST38|LARC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: Macrophage Inflammatory Protein 3 alpha
Clonality: monoclonal
Clone: 4N5F7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human