Anti-RBCK1 Rabbit Polyclonal Antibody
100 μg
Catalog # NOVUNBP2-27105
Supplier: Novus Biologicals
Jump to:
Product Details & Documents
Type: Primary
Antigen: RBCK1 (RanBP-type and C3HC4-type zinc finger containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Size:100 μg
- Western blot:Yes
- Cat. No.:NOVUNBP2-27105
- Clonality:Polyclonal
- Antibody type:Primary
- Gene ID:10616
- Antigen synonyms:XAP4|PBMEI|XAP3|HOIL-1|RBCK2|UBCE7IP3|HOIL1|ZRANB4|PGBM1|RNF54|C20orf18
- Storage temperature:Cool
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:RBCK1
- Shipping temperature:Ice
- Antigen name:RanBP-type and C3HC4-type zinc finger containing 1



