Anti-FHOD1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24748
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Formin homology 2 domain containing 1
- Antigen symbol:FHOD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Gene ID:29109
- Antigen synonyms:FHOS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: FHOD1 (formin homology 2 domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: