Anti-SH3GLB1 Mouse Monoclonal Antibody [clone: 30A882.1.1]
Catalog # NOVUNBP2-24733
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:SH3-domain GRB2-like endophilin B1
- Antigen symbol:SH3GLB1
- Clonality:Monoclonal
- Clone:30A882.1.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:51100
- Antigen synonyms:Bif-1|PPP1R70|CGI-61|dJ612B15.2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SH3GLB1 (SH3-domain GRB2-like endophilin B1)
Clonality: monoclonal
Clone: 30A882.1.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human