Anti-CYP1B1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24722
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:cytochrome P450, family 1, subfamily B, polypeptide 1
- Antigen symbol:CYP1B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:1545
- Antigen synonyms:CYPIB1|CP1B|P4501B1|GLC3A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CYP1B1 (cytochrome P450, family 1, subfamily B, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human