Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-00093B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Macrophage Scavenger Receptor 1
- Antigen symbol:MSR1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:4481
- Antigen synonyms:MRS-A|Scvr|SR-AII|Scara1|MSR|MSR-A|CD204|SR-AI|MSR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Macrophage Scavenger Receptor I
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse