Anti-GLI1 Rabbit Polyclonal Antibody (Biotin)
NOVUNB600-600B
- Antibody type:Primary
- Antigen name:GLI family zinc finger 1
- Antigen symbol:GLI1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- ChIP:Yes
- Gene ID:2735
- Antigen synonyms:Zfp5|GLI|Zfp-5|AV235269
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: GLI1 (GLI family zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse