Order Entry
Germany
ContactUsLinkComponent
Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB600-302
  • NOVUNB600-302SS
  • Antigen symbol:
  • c-Myc
  • c-Myc
  • Antigen name:
  • v-myc avian myelocytomatosis viral oncogene homolog
  • v-myc avian myelocytomatosis viral oncogene homolog
  • Antigen synonyms:
  • MRTL|bHLHe39|c-Myc|MYCC
  • MRTL|bHLHe39|c-Myc|MYCC
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 9E10
  • 9E10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4609
  • 4609
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together