Anti-ABCA1 Rat Monoclonal Antibody [clone: 5A1-1422.11]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ABCA1 (ATP-binding cassette, sub-family A (ABC1), member 1)
Clonality: monoclonal
Clone: 5A1-1422.11
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Specifications
- Catalog No:
- NOVUNB400-165SS
- NOVUNB400-165
- Antigen symbol:
- ABCA1
- ABCA1
- Antigen name:
- ATP Binding Cassette, sub-family A, member 1
- ATP Binding Cassette, sub-family A, member 1
- Antigen synonyms:
- ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
- ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 5A1-1422.11
- 5A1-1422.11
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 19
- 19
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice