Anti-ABCG1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB400-132G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily G, Member 1
- Antigen symbol:ABCG1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:9619
- Antigen synonyms:WHITE1,White protein homolog|MGC34313|WHT1|ABC8|ABCG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ABCG1 (ATP-binding cassette, sub-family G (WHITE), member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: