Anti-MMP2 Mouse Monoclonal Antibody [clone: 2C1]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: monoclonal
Clone: 2C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Guinea pig, Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNB200-113SS
- NOVUNB200-113
- Antigen symbol:
- MMP2
- MMP2
- Antigen name:
- Matrix Metalloproteinase 2
- Matrix Metalloproteinase 2
- Antigen synonyms:
- MMP-2|72 kDa gelatinase|matrix metalloproteinase-2|collagenase type IV-A|neutrophil gelatinase|gelatinase A|matrix metalloproteinase-II|72 kDa type IV collagenase
- MMP-2|72 kDa gelatinase|matrix metalloproteinase-2|collagenase type IV-A|neutrophil gelatinase|gelatinase A|matrix metalloproteinase-II|72 kDa type IV collagenase
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 2C1
- 2C1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 4313
- 4313
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice