Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB200-106G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 2A
- Antigen symbol:p19ARF
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:12578
- Antigen synonyms:CDKN2|p16-INK4|MLM|INK4|P16|p19ARF|p16-INK4a|p16INK4a|p16|CDKN2A|p16INK4|p19|p14|p16INK4A|CDK4I|INK4a|p14ARF|CMM2|ARF|MTS1|TP16
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse