Order Entry
Germany
ContactUsLinkComponent
Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: KEN]
Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: KEN]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: KEN]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-599SS
  • NOVUNB100-599
  • Antigen symbol:
  • BRCA1
  • BRCA1
  • Antigen name:
  • Breast Cancer type 1 susceptibility protein
  • Breast Cancer type 1 susceptibility protein
  • Antigen synonyms:
  • BROVCA1|BRCA1/BRCA2-containing complex|subunit 1|PPP1R53|breast and ovarian cancer sususceptibility protein 1|RING finger protein 53|IRIS|protein phosphatase 1|breast cancer type 1 susceptibility protein|RNF53|PNCA4|regulatory subunit 53|breast and ovarian cancer susceptibility protein 1|PSCP|BRCAI|BRCC1
  • BROVCA1|BRCA1/BRCA2-containing complex|subunit 1|PPP1R53|breast and ovarian cancer sususceptibility protein 1|RING finger protein 53|IRIS|protein phosphatase 1|breast cancer type 1 susceptibility protein|RNF53|PNCA4|regulatory subunit 53|breast and ovarian cancer susceptibility protein 1|PSCP|BRCAI|BRCC1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • KEN
  • KEN
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 672
  • 672
  • Isotype:
  • IgG3 kappa
  • IgG3 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information