Anti-PKR Rabbit Polyclonal Antibody
Catalog # BSBTPA2025
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 2 alpha kinase 2
- Antigen symbol:EIF2AK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:double-stranded RNA-activated protein kinase|Interferon-induced|Interferon-inducible RNA-dependent protein kinase|EIF2AK2|P1/eIF-2A protein kinase|PKR|p68 kinase
- Amino acid number:1 - 18
- Sequence:AGDLSAGFFMEELNTYR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human PKR (2-18aa AGDLSAGFFMEELNTYR).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Interferon-induced, double-stranded RNA-activated protein kinase (EIF2AK2) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: PKR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human