Anti-Annexin A2 Rabbit Polyclonal Antibody
BSBTPA1348
- Antibody type:Primary
- Antigen name:Annexin A2
- Antigen symbol:ANXA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:chromobindin 8|LIP2|LPC2|PAP-IV|annexin-2|protein I|placental anticoagulant protein IV|annexin II|LPC2D|CAL1H|ANX2L4|chromobindin-8|lipocortin II|calpactin I heavy chain|P36|calpactin I heavy polypeptide|calpactin-1 heavy chain|ANX2
- Amino acid number:121 - 141
- Sequence:RAEDGSVIDYELIDQDARDLY
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Annexin A2 (121-141aa RAEDGSVIDYELIDQDARDLY), different from the rat sequence by one amino acid
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Annexin A2 (ANXA2) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Annexin A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat