Anti-SMAD 1/2/3/5 Rabbit Polyclonal Antibody
Catalog # BSBTPA1331
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:SMAD 1/2/3/5
- Antigen symbol:SMAD 1/2/3/5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:442 - 456
- Sequence:GPLQWLDKVLTQMGS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human SMAD 1/2/3/5 (442-456aa GPLQWLDKVLTQMGS), identical to the related mouse and rat sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Mothers against decapentaplegic homolog 1 (SMAD1) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: SMAD 1/2/3/5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat