Order Entry
Germany
ContactUsLinkComponent
 
Anti-MTCO1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1317
Supplier:  Boster Bio
undefined
Anti-MTCO1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1317
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitochondrially Encoded Cytochrome C Oxidase I
  • Antigen symbol:
    MTCO1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Amino acid number:
    2 - 14
  • Sequence:
    FADRWLFSTNHKD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human MTCO1 (2-14aa FADRWLFSTNHKD)
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Cytochrome c oxidase subunit 1 (MT-CO1) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: MTCO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human