Order Entry
Germany
ContactUsLinkComponent
 
Anti-alpha-Tubulin Mouse Monoclonal Antibody [clone: Tub-1]
  BSBTMA1107
undefined
Anti-alpha-Tubulin Mouse Monoclonal Antibody [clone: Tub-1]
  BSBTMA1107

 

  • Antibody type:
    Primary
  • Antigen name:
    Tubulin alpha 1
  • Antigen symbol:
    TUBA1
  • Clonality:
    Monoclonal
  • Clone:
    Tub-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Chicken,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    tubulin alpha-4A chain|alpha 1|tubulin H2-alpha|tubulin|tubulin alpha-1 chain|H2-ALPHA
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Microtubules from chicken embryo brain
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Mouse IgG monoclonal antibody for alpha-Tubulin detection. Tested with WB, IHC-P in Human;rat. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: alpha-Tubulin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken