Anti-Vinculin Mouse Monoclonal Antibody [clone: VIN-54]
Catalog # BSBTMA1103
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Vinculin
- Antigen symbol:VINC
- Clonality:Monoclonal
- Clone:VIN-54
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:MVCL|CMH15|metavinculin|CMD1W
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human vinculin, purified from uterus
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for Vinculin, vinculin (VCL) detection. Tested with WB, IHC-F in Human;mouse;rat;chicken. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Vinculin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken