Anti-PI 3 Kinase catalytic subunit gamma/PI3K-gamma Mouse Monoclonal Antibody [clone: OTI4G10]
Catalog # ABCAAB140307-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:PI 3 Kinase catalytic subunit gamma
- Clonality:Monoclonal
- Clone:4G10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:African green monkey,Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PtdIns-3-kinase subunit gamma|Serine/threonine protein kinase PIK3CG|PI3-kinase subunit gamma|Phosphoinositide-3-kinase catalytic gamma polypeptide|PtdIns-3-kinase subunit p110-gamma|PI3Kgamma|PI3K-gamma|p110gamma|PIK3CG|p120-PI3K
- Storage buffer:pH: 7,3
Preservative: 0,02% Sodium azide
Constituents: PBS, 50% Glycerol (glycerin, glycerine), 1% BSA - Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:WB
ImmunoChemistry - Size:100 µl
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Mouse monoclonal [OTI4G10] to PI 3 Kinase catalytic subunit gamma/PI3K-gamma.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PI 3 Kinase catalytic subunit gamma/PI3K-gamma
Clonality: Monoclonal
Clone: OTI4G10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;African green monkey