Anti-6X His Tag Chicken Polyclonal Antibody
Catalog # ABCAAB134024-200
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:6X His Tag
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgY
- Western blot:Yes
- Form:Liquid
- Purity:Immunogen affinity purified
- Storage buffer:Preservative: 0,01% thimerosal (merthiolate)
Constituents: 49% PBS, 50% glycerol (glycerin, glycerine) - Storage temperature:Cool
- Concentration:200 µl, 1 mg/ml
- Shipping temperature:Ice
- Tested applications:WB
- Purification:IgY fractions were prepared from the yolks of eggs laid by immunised chickens
- Pk:200 µl
Specifications
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: 6X His Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: