Order Entry
Germany
ContactUsLinkComponent
Anti-NMDAR2A Rabbit Monoclonal Antibody [clone: EPR7063]
Anti-NMDAR2A Rabbit Monoclonal Antibody [clone: EPR7063]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-NMDAR2A Rabbit Monoclonal Antibody [clone: EPR7063]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB133265-100
  • ABCAAB133265-20UL
  • ABCAAB133265-1ML
  • Antigen symbol:
  • GRIN2A
  • Antigen name:
  • Glutamate receptor, ionotropic, N-methyl D-aspartate 2A
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR7063
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together