Anti-Lambda Light chain Rat Monoclonal Antibody [clone: JC5-1] (FITC (Fluorescein Isothiocyanate))
ABCAAB99623-250
: Abcam
- Antibody type:Secondary
- Antigen name:lambda light chain
- Clonality:Monoclonal
- Clone:JC5-1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Seq:1370
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pack type:Vial
- Pk:250 µG
Rat monoclonal [JC5-1] Anti-Mouse lambda light chain (FITC).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Secondary
Antigen: Lambda Light chain
Clonality: Monoclonal
Clone: JC5-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse