Order Entry
Germany
ContactUsLinkComponent
 
Anti-Digoxigenin Chicken Polyclonal Antibody
  ABCAAB51946-200
 :  Abcam
Anti-Digoxigenin Chicken Polyclonal Antibody
  ABCAAB51946-200
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Digoxigenin
  • Antigen symbol:
    DIG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Chicken
  • Isotype:
    IgY
  • Western blot:
    Yes
  • Seq:
    1358
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µg
  • Pack type:
    Vial
  • Pk:
    200 µG

 

 

Chicken polyclonal to Digoxigenin.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Digoxigenin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: