Anti-CDK1+CDK2+CDK3+CDK5 Rabbit Monoclonal Antibody [clone: E53]
: Abcam
New Product
:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: monoclonal
Clone: E53
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- ABCAAB32030-100
- ABCAAB32030-1ML
- Antigen symbol:
- CDK1
- Antigen name:
- Cyclin-dependent kinase 1
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- E53
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoPrecipitation:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice