Order Entry
Germany
ContactUsLinkComponent
Anti-GAPDH Rabbit Polyclonal Antibody
Anti-GAPDH Rabbit Polyclonal Antibody
Supplier:  Abcam
Certificates
Anti-GAPDH Rabbit Polyclonal Antibody
Supplier:  Abcam

Specifications

  • Catalog No:
  • ABCAAB9485-100
  • Antigen symbol:
  • GAPDH
  • Antigen name:
  • Glyceraldehyde-3-phosphate dehydrogenase
  • Antibody type:
  • Primary
  • Clonality:
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Host:
  • Rabbit
  • Western blot:
  • Yes
  • ELISA:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications