Order Entry
Germany
ContactUsLinkComponent
Anti-68kDa Neurofilament/NF-L Mouse Monoclonal Antibody [clone: DA2]
Anti-68kDa Neurofilament/NF-L Mouse Monoclonal Antibody [clone: DA2]
Catalog # ABCAAB7255-250
Supplier:  Abcam
undefined
Anti-68kDa Neurofilament/NF-L Mouse Monoclonal Antibody [clone: DA2]
Catalog # ABCAAB7255-250
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurofilament, Light Polypeptide
  • Antigen symbol:
    NEFL
  • Clonality:
    Monoclonal
  • Clone:
    DA2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 μl
  • Pack type:
    Vial
  • Pk:
    250 µl

Specifications

About this item

Mouse monoclonal [DA2] to 68kDa Neurofilament/NF-L.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: 68kDa Neurofilament/NF-L
Clonality: Monoclonal
Clone: DA2
Conjugation:
Epitope: Epitope mapped to a short peptide in the C-terminal “tail” region of the molecule within the human sequence SYYTSHVQEEQIEVE, amino acids 441-455
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse