Anti-OLFM3 Mouse Polyclonal Antibody
Catalog # ABNOH00118427-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Olfactomedin 3
- Antigen symbol:OLFM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:118427
- Antigen synonyms:Olfactomedin-3|Olfm3|Olfactomedin related ER localized protein 3|NOELIN3 V2|NOE3_HUMAN|NOELIN3 V6|NOELIN3 V4|Olfactomedin 3|NOELIN3|NOELIN3 V3|Noe3|Olfactomedin3|NOELIN3 V1|Noelin-3|NOELIN3 V5|Noelin 3|Optimedin.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: OLFM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human