Anti-CAB39L Mouse Monoclonal Antibody [clone: 3B11-1A4]
Catalog # ABNOH00081617-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Calcium Binding Protein 39-like
- Antigen symbol:CAB39L
- Clonality:Monoclonal
- Clone:3B11-1A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:81617
- Antigen synonyms:MO2L|MO25-BETA|bA103J18.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CAB39L
Clonality: Monoclonal
Clone: 3B11-1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human