Order Entry
Germany
ContactUsLinkComponent
Anti-DBR1 Mouse Monoclonal Antibody [clone: 3A7]
Anti-DBR1 Mouse Monoclonal Antibody [clone: 3A7]
  ABNOH00051163-M01
 :  Abnova
undefined
Anti-DBR1 Mouse Monoclonal Antibody [clone: 3A7]
  ABNOH00051163-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    debranching RNA lariats 1
  • Antigen symbol:
    DBR1
  • Clonality:
    Monoclonal
  • Clone:
    3A7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    51163
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: DBR1
Clonality: Monoclonal
Clone: 3A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human