Anti-GMPPB Mouse Polyclonal Antibody
ABNOH00029925-A01
: Abnova
- Antibody type:Primary
- Antigen name:GDP Mannose Pyrophosphorylase B
- Antigen symbol:GMPPB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:29925
- Antigen synonyms:GTP-mannose-1-phosphate guanylyltransferase beta|KIAA1851|GMPPB|Mannose-1-phosphate guanyltransferase beta.|GMPPB_HUMAN|GDP mannose pyrophosphorylase B|GDP-mannose pyrophosphorylase B|Mannose 1 phosphate guanylyltransferase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: GMPPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human