Anti-TAGLN3 Mouse Monoclonal Antibody [clone: 1D2]
Catalog # ABNOH00029114-M01
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Transgelin 3
- Antigen-Symbol:TAGLN3
- Klonalität:Monoklonal
- Klon:1D2
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:No
- Gen-ID:29114
- Antigen-Synonyme:NP25|NP22|NP24
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µg
- VE:100 µg
Specifications
About this item
Type: Primary
Antigen: TAGLN3
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human