Anti-HEY2 Mouse Monoclonal Antibody [clone: 2B10]
ABNOH00023493-M02
: Abnova
- Antibody type:Primary
- Antigen name:Hairy/enhancer-of-split related with YRPW motif 2
- Antigen symbol:HEY2
- Clonality:Monoclonal
- Clone:2B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23493
- Antigen synonyms:GRL|bHLHb32|GRIDLOCK|HESR2|HRT2|HERP1|CHF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: HEY2
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human