Anti-SLC4A1AP Mouse Monoclonal Antibody [clone: 2G10]
ABNOH00022950-M01
: Abnova
- Antibody type:Primary
- Antigen name:Solute Carrier Family 4 (anion exchanger), Member 1, Adaptor Protein
- Antigen symbol:Solute carrier family 4, member 1AP
- Clonality:Monoclonal
- Clone:2G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:22950
- Antigen synonyms:HLC3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SLC4A1AP
Clonality: Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human