Anti-TFEC Mouse Monoclonal Antibody [clone: 4F11]
Catalog # ABNOH00022797-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor EC
- Antigen symbol:TFEC
- Clonality:Monoclonal
- Clone:4F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:22797
- Antigen synonyms:TFE-C|bHLHe34|TCFEC|hTFEC-L|TFEC-L|TFECL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TFEC
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human