Anti-CCS Mouse Monoclonal Antibody [clone: 1E2]
ABNOH00009973-M03
: Abnova
- Antibody type:Primary
- Antigen name:Copper Chaperone for Superoxide Dismutase
- Antigen symbol:CCS
- Clonality:Monoclonal
- Clone:1E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9973
- Antigen synonyms:CCS|copper chaperone for superoxide dismutase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CCS
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human