Anti-OXSR1 Mouse Monoclonal Antibody [clone: 2A2-1A2]
Catalog # ABNOH00009943-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Oxidative Stress Responsive 1
- Antigen symbol:OXSR1
- Clonality:Monoclonal
- Clone:2A2-1A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9943
- Antigen synonyms:OXSR 1|KIAA1101|Oxidative stress responsive 1 protein|OSR1|OSR 1|Serine/threonine protein kinase OSR1.|Oxidative stress responsive 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: OXSR1
Clonality: Monoclonal
Clone: 2A2-1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human