Anti-CTIF Mouse Polyclonal Antibody
Catalog # ABNOH00009811-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CBP80/20-dependent translation initiation factor
- Antigen symbol:CTIF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9811
- Antigen synonyms:KIAA0427|Gm672
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CTIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human