Order Entry
Germany
ContactUsLinkComponent
Anti-DOCK4 Mouse Monoclonal Antibody [clone: 3E7]
Anti-DOCK4 Mouse Monoclonal Antibody [clone: 3E7]
Catalog # ABNOH00009732-M01A
Supplier:  Abnova
undefined
Anti-DOCK4 Mouse Monoclonal Antibody [clone: 3E7]
Catalog # ABNOH00009732-M01A
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 4
  • Antigen symbol:
    DOCK4
  • Clonality:
    Monoclonal
  • Clone:
    3E7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9732
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: DOCK4
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human