Order Entry
Germany
ContactUsLinkComponent
Anti-HOMER1 Rabbit Polyclonal Antibody
Anti-HOMER1 Rabbit Polyclonal Antibody
Catalog # ABNOH00009456-D01P
Supplier:  Abnova
undefined
Anti-HOMER1 Rabbit Polyclonal Antibody
Catalog # ABNOH00009456-D01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Homer protein homolog 1
  • Antigen symbol:
    HOMER1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9456
  • Antigen synonyms:
    HOMER|Ves-1|SYN47|HOMER1B|HOMER1A|HOMER1C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: HOMER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse