Anti-MED7 Mouse Monoclonal Antibody [clone: 3E7]
Catalog # ABNOH00009443-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 7
- Antigen symbol:MED7
- Clonality:Monoclonal
- Clone:3E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:9443
- Antigen synonyms:CRSP9|CRSP33|ARC34
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MED7
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human