Order Entry
Germany
ContactUsLinkComponent
Anti-NOLC1 Mouse Monoclonal Antibody [clone: 6B4]
Anti-NOLC1 Mouse Monoclonal Antibody [clone: 6B4]
Catalog # ABNOH00009221-M02
Supplier:  Abnova
undefined
Anti-NOLC1 Mouse Monoclonal Antibody [clone: 6B4]
Catalog # ABNOH00009221-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nucleolar and coiled-body phosphoprotein 1
  • Antigen symbol:
    NOLC1
  • Clonality:
    Monoclonal
  • Clone:
    6B4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9221
  • Antigen synonyms:
    NOPP130|nucleolar and coiled-body phosphoprotein 1|NOPP140|KIAA0035|P130|nucleolar phosphoprotein p130|nucleolar protein p130|HCV NS5A trans-regulated protein 13|nucleolar and coiled-body phosphprotein 1|NS5ATP13
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: NOLC1
Clonality: Monoclonal
Clone: 6B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human