Anti-STBD1 Mouse Monoclonal Antibody [clone: 4H4]
Catalog # ABNOH00008987-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Starch binding domain 1
- Antigen symbol:STBD1
- Clonality:Monoclonal
- Clone:4H4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8987
- Antigen synonyms:GENX-3414|Glycophagy Cargo Receptor STBD1|Genethonin 1|GENEX3414
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: STBD1
Clonality: Monoclonal
Clone: 4H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human