Anti-USO1 Mouse Monoclonal Antibody [clone: 3F4]
Catalog # ABNOH00008615-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:USO1 vesicle transport factor
- Antigen symbol:USO1
- Clonality:Monoclonal
- Clone:3F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8615
- Antigen synonyms:VDP|P115|TAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: USO1
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human